Order Entry
Netherlands
ContactUsLinkComponent
Petroleum spirit, 40…65 °C, Technical Grade
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…65 °C, Technical Grade
Catalog # ACRO444290050
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…65 °C, Technical Grade
Catalog # ACRO444290050
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Boiling point:
    40 to 65°C (760 mmHg)
  • Density:
    (15°C) 0.645 to 0.665 g/ml
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %

Specification Test Results

About this item

 :
Boiling Pt:40…65 °C (760 mmHg)
Density:0.65 g/cm³
Flash Pt:–55 °C
Storage Temperature:Refrigerator
UN:3295
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:64742-49-0
 :8032-32-4
MDL Number:MFCD00081849