
1-Methyl-1H-pyrazole-4-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5952-92-1
:
Formula:C₅H₆N₂O₂
MDL Number:MFCD00159641
:126.1100 g/mol
Melting Pt:207…209 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together