
2-Methoxy-4-nitrobenzonitrile 98%
Catalog # ACRO440220050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to brown Powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:178.15 g/mol
Melting Pt:179…181 °C
ADR:6.1,III
:GHS07
Storage Temperature:Ambient
CAS Number:101084-96-2
Formula:C₈H₆N₂O₃
MDL Number:MFCD02093781