Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
N-Phenyl-2-naphthylamine 97%
 
undefined
 
 
 
N-Phenyl-2-naphthylamine 97%

 

  • Appearance:
    Off-white to grey to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    105 to 108 °C
  • HPLC:
    ≥96.0 %
  • Titration with HClO4:
    ≥96.0 %

 

 

Frequently Bought Together