Order Entry
Netherlands
ContactUsLinkComponent
Lithium acetylide ethylenediamine complex 25% (w/w) slurry in toluene, AcroSeal®
Flammable
Corrosive
Irritant
Health
Lithium acetylide ethylenediamine complex 25% (w/w) slurry in toluene, AcroSeal®
Catalog # 439881000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Lithium acetylide ethylenediamine complex 25% (w/w) slurry in toluene, AcroSeal®
Catalog # 439881000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Tan to orange-brown Slurry
  • Titration with HClO4:
    23.5 to 30.0 %

Specification Test Results

About this item

 :
 :92.07 g/mol
Flash Pt:–14 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:6867-30-7
Formula:C₄H₉LiN₂
MDL Number:MFCD00013183