
5,5-Dibromopyrimidine-2,4,6(1H,3H,5H)-trione 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:511-67-1
:
Formula:C₄H₂Br₂N₂O₃
MDL Number:MFCD00010595
Melting Pt:240…242 °C
UN:3261
ADR:8,II
:285.89 g/mol
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together