Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-5-methoxybenzyl bromide 97%
Corrosive
2-Bromo-5-methoxybenzyl bromide 97%
Catalog # ACRO439790250
CAS Number:  
undefined
Corrosive
2-Bromo-5-methoxybenzyl bromide 97%
Catalog # ACRO439790250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    90 to 94 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :279.96 g/mol
Melting Pt:91…93 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:19614-12-1
Formula:C₈H₈Br₂O
MDL Number:MFCD00078664