
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1421-65-4
:
Formula:C₁₀H₁₄ClNO₄
MDL Number:MFCD00038958
Melting Pt:94.2…94.3 °C
:247.675 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light brown Powder
- Infrared spectrum:Conforms
- Water:≤5.00 %
- Elemental Analysis:45.58 to 51.40 % (C) On dry basis
- Specific optical rotation:+9.0° to +11.0° (20°C, 589 nm) (C=2 in methanol)
Specification Test Results
Frequently Bought Together