Order Entry
Netherlands
ContactUsLinkComponent
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%
Irritant
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%
Certificates
undefined
Irritant
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    49 to 53 °C
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)

Specification Test Results

Related Information

Frequently Bought Together