
1,1'-Bis((2S,5S)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:854920-95-9
:
Formula:C₃₈H₆₀BF₄FeP₂Rh⁻
MDL Number:MFCD15144872
:824.4 g/mol
Melting Pt:235…238 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- NMR:≥96.0 %
Specification Test Results
Frequently Bought Together