



Rhodium(III) chloride hydrate 99.95% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20765-98-4
:
Formula:Cl₃Rh·H₂O
MDL Number:MFCD00149839
:227.28 g/mol
Melting Pt:100 °C (dec.)
Storage Temperature:Ambient
UN:3260
ADR:8,III
:GHS07
:GHS05
:GHS08
:GHS09
Specification Test Results
- Appearance:Red-brown to brown to black Crystals
- Assay:37.0 to 43.0 % (Rh)
- Total trace metal impurities:≤500 ppm
Specification Test Results
Frequently Bought Together