Order Entry
Netherlands
ContactUsLinkComponent
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Certificates
undefined
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Certificates

Specification Test Results

  • Appearance:
    Colorless to white Crystals
  • Total trace metal impurities:
    ≤10 ppm
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together