Poly(DL-lactide-co-glycolide), M.W. ∼ 19,000 ( Lactide:Glycolide) (50:50)%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26780-50-7
:
Formula:[C₃H₄O₂]x[C₂H₂O₂]y
MDL Number:MFCD00131930
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to tan Crystalline powder or granules or chunks
- Viscosity:0.15 to 0.25 dl/g
- Polymer:45 to 55 % (Lactide)
Specification Test Results
Frequently Bought Together