2'-Bromoacetanilide 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:614-76-6
:
Formula:H₃CC(O)NH(C₆H₄Br)
MDL Number:MFCD00099252
:214.06 g/mol
Melting Pt:100…102 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:98 to 103 °C
- GC:≥95.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together