Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Irritant
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Catalog # ACRO435182500
CAS Number:  
undefined
Irritant
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Catalog # ACRO435182500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Orange to red to brown Powder
  • Infrared spectrum:
    Conforms
  • NMR (1H-NMR):
    Consistent with structure
  • NMR (19F-NMR):
    Consistent with structure
  • NMR (31P-NMR):
    Consistent with structure
  • Elemental Analysis:
    52.4 to 56.8 % C
  • Specific optical rotation:
    +40° to +50° (20°C, 589 nm) (c=1 in methanol)

Specification Test Results

About this item

 :
 :660.371 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:228121-39-9
Formula:C₃₀H₄₈BF₄P₂Rh
MDL Number:MFCD01862464