
Ethyl-2-cyanoacrylate
Catalog # ACRO434450050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- NMR:Consistent with structure
- NMR (1H-NMR):Consistent with structure
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
:125.1253 g/mol
Boiling Pt:54…56 °C (760 mmHg)
Melting Pt:–20 °C
Density:1.04 g/cm³
Flash Pt:85 °C
Storage Temperature:Freezer
UN:3335
ADR:9,III
:GHS07
CAS Number:7085-85-0
Formula:C₆H₇NO₂
MDL Number:MFCD00045615