Order Entry
Netherlands
ContactUsLinkComponent
4-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
 
 
 
 
4-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
  433690500.
 :  
undefined
 
 
 
 
4-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
  433690500.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

 

 

  • Appearance:
    Yellow to brown Solution
  • Titration Grignard:
    0.22 to 0.28 M
  • Water:
    ≤3.0 %
  • pH:
    6.0 to 8.0 (10 % in water)
  • Hydroxyl value:
    65 to 78 mg KOH/g
  • Iodine value:
    25 to 35 g I2/100 g
  • Saponification value:
    65 to 70 mg KOH/g

 

 

 :
 :245.402 g/mol
 :0.948 g/cm³
 :–17 °C
 :Ambient
 :2924
 :3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :115913-58-1
 :C₁₁H₉BrMg
 :MFCD01311487