



2-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 433680500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.22 to 0.28 M
- Additional info:May darken on storage
- GC:≥94 %
Specification Test Results
About this item
:
:245.402 g/mol
Boiling Pt:65 °C (760 mmHg)
Density:0.921 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS07
:GHS05
:GHS08
CAS Number:21450-90-8
Formula:C₁₁H₉BrMg
MDL Number:MFCD01311486