



3,5-Dimethyl-4-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
431960500.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
- Appearance:Yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- Refractive index:1.5830 to 1.5850 (20°C, 589 nm)
- GC:≥97.0 %
:
:239.395 g/mol
:0.949 g/cm³
:–17 °C
:Ambient
:2924
:3,II
:GHS07
:GHS05
:GHS02
:GHS08
:185416-17-5
:C₉H₁₁BrMgO
:MFCD01319917