



4-Biphenylmagnesium bromide 0.5 M in THF, AcroSeal®
431621000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- HPLC:≥97.5 %
- Titration with NaOH:≥97.5 %
:
:257.413 g/mol
:65 °C (760 mmHg)
:0.961 g/cm³
:–17 °C
:Ambient
:2924
:3,II
:GHS07
:GHS05
:GHS02
:GHS08
:3315-91-1
:C₁₂H₉BrMg
:MFCD01311479