Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,3-Benzothiazole-6-carboxaldehyde 97%
 
undefined
 
1,3-Benzothiazole-6-carboxaldehyde 97%

 

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    90 to 96 °C
  • HPLC:
    ≥96.0 %

 

 

Frequently Bought Together