
2-Chloro-4-fluorophenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:313545-72-1
:
Formula:C₆H₅BClFO₂
MDL Number:MFCD02684295
:174.37 g/mol
Melting Pt:228…230 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together