
3-Bromo-4-chlorobenzoic acid 97%
Catalog # ACRO429870050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:219 to 223 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:235.46 g/mol
Melting Pt:220…222 °C
:GHS07
Storage Temperature:Ambient
CAS Number:42860-10-6
Formula:C₇H₄BrClO₂
MDL Number:MFCD00079706