
2-Chloropyrimidine-5-sulphonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98026-88-1
:
Formula:C₄H₂Cl₂N₂O₂S
MDL Number:MFCD06657208
:213.04 g/mol
Melting Pt:64…67 °C
Storage Temperature:Freezer
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to yellow to light orange Crystalline solid
- Infrared spectrum:Conforms
- Melting point:64 to 68 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together