Order Entry
Netherlands
ContactUsLinkComponent
Lithium tetrachlorocuprate 0.1M in THF, AcroSeal®
Irritant
Flammable
Health
Lithium tetrachlorocuprate 0.1M in THF, AcroSeal®
Catalog # 429051000.
CAS Number:  
undefined
Irritant
Flammable
Health
Lithium tetrachlorocuprate 0.1M in THF, AcroSeal®
Catalog # 429051000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear orange to red-brown Solution
  • Titration Argentometric:
    0.09 to 0.12 M

Specification Test Results

About this item

 :
 :219.24 g/mol
Density:0.91 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
 :GHS07
 :GHS02
 :GHS08
CAS Number:15489-27-7
Formula:Cl₄CuLi₂
MDL Number:MFCD00011081