

Sodium trimethylsilanolate 1 M in dichloromethane, AcroSeal®
Catalog # 428841000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to amber Solution (May develop some turbidity or precipitate)
- Titration with HClO4:0.90 to 1.10 M
Specification Test Results
About this item
:
:112.179 g/mol
Density:1.28 g/cm³
Flash Pt:63 °C
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS08
:GHS05
CAS Number:18027-10-6
Formula:C₃H₉NaOSi
MDL Number:MFCD00013926