Order Entry
Netherlands
ContactUsLinkComponent
 
Castor oil ethoxylated pH 6.0-8.1
 
undefined
Castor oil ethoxylated pH 6.0-8.1

 

  • Appearance:
    White to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤3.0 %
  • pH:
    6.0 to 8.0 (10 % in water)
  • Hydroxyl value:
    65 to 78 mg KOH/g
  • Iodine value:
    25 to 35 g I2/100 g
  • Saponification value:
    65 to 70 mg KOH/g

 

 

Frequently Bought Together