

Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-96-1
:
Formula:C₈H₁₈O
MDL Number:MFCD00009461
:130.23 g/mol
Boiling Pt:141 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.768 g/cm³
Flash Pt:25 °C
Storage Temperature:Ambient
UN:1149
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.002 %
Specification Test Results
Related Information
Frequently Bought Together