Order Entry
Netherlands
ContactUsLinkComponent
Ammonia 0.5 M in THF, AcroSeal®
Irritant
Flammable
Health
Ammonia 0.5 M in THF, AcroSeal®
Catalog # 388441000.
CAS Number:  
undefined
Irritant
Flammable
Health
Ammonia 0.5 M in THF, AcroSeal®
Catalog # 388441000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Water:
    ≤2 % (K.F.)
  • Strength:
    0.40 to 0.60 M

Specification Test Results

About this item

 :
 :17.03 g/mol
Density:0.85 g/cm³
Flash Pt:–36 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
 :GHS07
 :GHS02
 :GHS08
CAS Number:7664-41-7
Formula:NH₃
MDL Number:MFCD00011418