



4-Chlorophenylmagnesium bromide 1 M in THF/toluene, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:873-77-8
:
Formula:C₆H₄BrClMg
MDL Number:MFCD00009926
:215.76 g/mol
Density:1 g/cm³
Flash Pt:–21 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to green to brown Slightly hazy solution
- Titration Active base:21.0 to 25.0 %
Specification Test Results
Related Information
Frequently Bought Together