
2-Propanesulphonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10147-37-2
:
Formula:C₃H₇ClO₂S
MDL Number:MFCD00007453
:142.61 g/mol
Boiling Pt:95 °C (45 mmHg)
Melting Pt:–47 °C
Density:1.273 g/cm³
Flash Pt:86 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4580 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together