Boranepyridine complex 95%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Pyridine Borane, Borane, pyridine complex
- CAS Number:110-51-0
- Formula:C₅H₈BN
- MDL Number:MFCD00012435
- Molecular Weight:92.94 g/mol
- Melting Pt:10…11 °C
- Density:0.92 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerator
- UN:2810
- ADR:6.1,II
2 Options Available Below
Product Details & Documents
Synonyms: Pyridine Borane, Borane, pyridine complex
- CAS Number:110-51-0
- Formula:C₅H₈BN
- MDL Number:MFCD00012435
- Molecular Weight:92.94 g/mol
- Melting Pt:10…11 °C
- Density:0.92 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerator
- UN:2810
- ADR:6.1,II
Documents
ACRO17701
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear to slightly hazy colorless to amber Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Titration:≥94 % (Active hydrogen)
- HPLC:≥95.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



