Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Corrosive
Flammable
o-Tolylmagnesium Bromide 2 M in diethyl ether, AcroSeal®
Catalog # 427471000.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
o-Tolylmagnesium Bromide 2 M in diethyl ether, AcroSeal®
Catalog # 427471000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Grey to brown Solution
  • Titration Active base:
    1.90 to 2.20 M
  • Melting point:
    184 to 187 °C
  • HPLC:
    ≥98.0 %

Specification Test Results

About this item

 :
 :195.34 g/mol
Density:1.013 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:2924
ADR:3,I
 :GHS07
 :GHS05
 :GHS02
CAS Number:932-31-0
Formula:C₇H₇BrMg
MDL Number:MFCD00010350