
4-(tert-Butyldimethylsilyloxy)cyclohexanone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55145-45-4
:
Formula:C₁₂H₂₄O₂Si
MDL Number:MFCD06411307
:228.407 g/mol
Boiling Pt:85 °C (1 mmHg)
Density:0.909 g/cm³
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together