



Phenethylmagnesium chloride solution 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90878-19-6
:
Formula:C₈H₉ClMg
MDL Number:MFCD00192387
:164.915 g/mol
Density:0.954 g/cm³
Flash Pt:–17 °C
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.90 to 1.10 M
- Refractive index:1.3820 to 1.3940 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Related Information
Frequently Bought Together