
Methyl-1-cyclopentene-1-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25662-28-6
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00239506
:126.1531 g/mol
Boiling Pt:76…78 °C (9 mmHg)
Flash Pt:62 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together