Order Entry
Netherlands
ContactUsLinkComponent
1,1-Diphenylethylene oxide
Irritant
Flammable
1,1-Diphenylethylene oxide
Certificates
undefined
Irritant
Flammable
1,1-Diphenylethylene oxide

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    52 to 57 °C
  • NMR:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together