Order Entry
Netherlands
ContactUsLinkComponent
Isobutylamine
Flammable
Corrosive
Toxic
Isobutylamine
Certificates
undefined
Flammable
Corrosive
Toxic
Isobutylamine

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3960 to 1.3980 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Additional info:
    is possible)

Specification Test Results

Related Information

Frequently Bought Together