(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 95%, 98% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:261165-05-3
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD01320857
:229.28 g/mol
Melting Pt:111.8 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder and/or chunks
- Infrared spectrum:Conforms
- HPLC:≥94 % (ELSD)
- Enantiomeric excess:≥97.5 % (HPLC)
Specification Test Results
Frequently Bought Together