Order Entry
Netherlands
ContactUsLinkComponent
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
Toxic
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
Catalog # ACRO356520010
CAS Number:  
undefined
Toxic
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
Catalog # ACRO356520010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    -149.5° to -155.5° (20°C, 589 nm) (c=1, pyridine)
  • Titration with HClO4:
    96.0 to 104.0 %

Specification Test Results

About this item

CAS Number:18531-95-8
 :
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00145204
 :284.362 g/mol
Melting Pt:242…244 °C
 :GHS06
Storage Temperature:Ambient