Order Entry
Netherlands
ContactUsLinkComponent
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 99%
Irritant
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 99%
Catalog # ACRO356510010
CAS Number:  
undefined
Irritant
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 99%
Catalog # ACRO356510010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Off-white to brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • Specific optical rotation:
    +153° to +159° (20°C, 589 nm) (c=1, pyridine)

Specification Test Results

About this item

CAS Number:18741-85-0
 :
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00145204
 :284.362 g/mol
Melting Pt:242…244 °C
 :GHS07
Storage Temperature:Ambient