Phorbol-12-myristate-13-acetate 97%
: TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate
:16561-29-8
:
:C₃₆H₅₆O₈
:MFCD00036736
:616.836 g/mol
:72 °C
:GHS07
:Freezer
- Environmentally Preferable:
- Appearance:Colorless to white to light yellow Film or foam
- HPLC:≥96.0 %
Frequently Bought Together