
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98014-14-3
:
Formula:C₂₀H₁₈Cl₂N₄ORu
MDL Number:MFCD00150005
:502.361 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green to black Powder and/or chunks
- Infrared spectrum:Conforms
- Assay:≥96.0 % (Based on Carbon)
Specification Test Results
Frequently Bought Together