
2-Methyl-5-nitroimidazole-1-ethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:443-48-1
:
Formula:C₆H₉N₃O₃
MDL Number:MFCD00009750
:171.16 g/mol
Melting Pt:159…163 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:159 to 163 °C
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together