
(R)-(−)-Phenylephrine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61-76-7
:
Formula:C₉H₁₃NO₂·HCl
MDL Number:MFCD00012605
Melting Pt:140…145 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:140 to 145 °C
- Loss on drying:≤1 % (1 g, 105 °C)
- Specific optical rotation:-43° to -47° (20 °C, 589 nm) (c=2, H2O) (on dried substance)
- Titration with HClO4:≥98.5 % (On dried substance)
Specification Test Results
Frequently Bought Together