Leuco crystal violet 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:603-48-5
:
Formula:C₂₅H₃₁N₃
MDL Number:MFCD00008314
:373.54 g/mol
Melting Pt:175…180 °C
UN:3077
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light grey to light violet Powder
- Infrared spectrum:Conforms
- Melting point:175 to 180 °C
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together