Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
Lithium acetylide ethylenediamine complex 85%
 
undefined
 
 
 
Lithium acetylide ethylenediamine complex 85%

 

  • Appearance:
    Light yellow to light brown Crystalline powder or crystals
  • Assay:
    ≥80 % (Gasvolumetric)
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥97.5 %

 

 

Frequently Bought Together