Order Entry
Netherlands
ContactUsLinkComponent
DL-Valinol 97%
Irritant
DL-Valinol 97%
Catalog # ACRO180420050
CAS Number:  
undefined
Irritant
DL-Valinol 97%
Catalog # ACRO180420050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4570 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

CAS Number:16369-05-4
 :
Formula:C₅H₁₃NO
MDL Number:MFCD00004730
 :103.17 g/mol
Boiling Pt:75…77 °C (8 mmHg)
Density:0.93 g/cm³
Flash Pt:90 °C
 :GHS07
Storage Temperature:Refrigerator