5-Nitro-2-furaldehyde diacetate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-55-7
:
Formula:C₉H₉NO₇
MDL Number:MFCD00003244
:243.17 g/mol
Melting Pt:89…93 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:89 to 93 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together