Tolazoline hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59-97-2
:
Formula:C₁₀H₁₃ClN₂
MDL Number:MFCD00012693
Molecular Weight:196.678 g/mol
Melting Pt:172…176 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:172 to 176 °C
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together